Tisk…

Dimethyl-3,4-dihydroxypyrrole-2,5-dicarboxylate 97%

Dodavatel: Thermo Scientific

A19238.03 A19238.06
A19238.03EA 934 CZK
A19238.03 A19238.06
Dimethyl-3,4-dihydroxypyrrole-2,5-dicarboxylate 97%
Dimethyl-3,4-dihydroxypyrrole-2,5-dicarboxylate

Purity: 97% CAS No.: 1632-19-5 Molecular Formula: C8H9NO6 Formula Weight: 215.16 Flash Point: 110°(230°F) Melting Point: 209-212° Boiling Point: 268-272° MDL No.: MFCD00276587

Vzorec: C₈H₉NO₆
MW: 215,16 g/mol
Bod varu: 268…272 °C
Bod tání: 209…212 °C
Hustota: 1,248
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00276587
CAS číslo: 1632-19-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR