(+)-Nootkatone ≥98%, crystalline
Dodavatel: Thermo Scientific
Synonyms:
(4R,4aS,6R)-4,4a-Dimethyl-6-(prop-1-en-2-yl)-4,4a,5,6,7,8-hexahydronaphthalen-2(3H)-one
A19166.03
A19166.06
A19166.14
A19166.03EA
1960
CZK
A19166.03
A19166.06
A19166.14
(+)-Nootkatone ≥98%, crystalline
(+)-Nootkatone
Purity: 98+%
CAS No.: 4674-50-4
Molecular Formula: C15H22O
Formula Weight: 218.34
Storage Temperature: 2°C to 8°C
Flash Point: 99°(210°F)
Melting Point: 34-36°
Boiling Point: 125°/0.5mm
Beilstein Reference: 4676969
Optical Rotation: +185° (c=1 in chloroform)
MDL No.: MFCD00036591
Vzorec:
C₁₅H₂₂O MW: 218,34 g/mol Bod varu: 125 °C (0,5 mmHg) Bod tání: 34…36 °C Hustota: 0,997 Bod vzplanutí: 99 °C (210 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD04974578 CAS číslo: 4674-50-4 EINECS: 225-124-4 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...