5-Acetylindan 97%
Dodavatel: Thermo Fisher Scientific
A19102.09
A19102.18
A19102.88
A19102.09EA
5430
CZK
A19102.09
A19102.18
A19102.88
5-Acetylindan 97%
5-Acetylindan
Purity: 97%
CAS No.: 4228-10-8
Molecular Formula: C11H12O
Formula Weight: 160.22
Boiling Point: 85-87°/0.05mm
MDL No.: MFCD00021246
Vzorec:
C₁₁H₁₂O Bod varu: 85…87 °C (0,05 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00021246 CAS číslo: 4228-10-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...