Boc-ON ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-(tert-BOC-Oxyimino)-2-phenylacetonitrile, N-((tert-Butoxycarbonyl)oxy)benzimidoyl cyanide, 2-(Boc-oxyimino)-2-phenylacetonitrile, 2-(tert-Butoxycarbonyloxyimino)-2-phenylacetonitrile, [(cyano-phenyl-methylidene)amino] tert-butyl carbonate
A18906.06
A18906.14
A18906.22
A18906.06EA
1090
CZK
A18906.06
A18906.14
A18906.22
Boc-ON ≥98%
Boc-ON
Purity: 98+%
CAS No.: 58632-95-4
Molecular Formula: C13H14N2O3
Formula Weight: 246.27
Storage Temperature: 2°C to 8°C
Melting Point: 86-89°
Beilstein Reference: 2217296
UN No.: UN3439
MDL No.: MFCD00001863
Vzorec:
C₁₃H₁₄N₂O₃ MW: 246,27 g/mol Bod tání: 86…89 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001863 CAS číslo: 58632-95-4 EINECS: 261-370-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...