Tisk…

Geranic acid (sum of isomers) 98%

Dodavatel: Thermo Scientific
Varování

A18750.14 A18750.22
A18750.14EA 1920 CZK
A18750.14 A18750.22
Geranic acid (sum of isomers) 98%
Geranic acid (sum of isomers)

Purity: 90+% CAS No.: 459-80-3 Molecular Formula: C10H16O2 Formula Weight: 168.24 Flash Point: 133°(271°F) Refractive Index: 1.4840 Boiling Point: 249-251° Beilstein Reference: 1763804 MDL No.: MFCD00036493

Vzorec: C₁₀H₁₆O₂
MW: 168,24 g/mol
Bod varu: 249…251 °C
Hustota: 0,955
Bod vzplanutí: 133 °C (271 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036493
CAS číslo: 459-80-3
EINECS: 207-299-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR