Tisk…

β-Alanine benzyl ester p-toluenesulphonate 98%

Dodavatel: Thermo Scientific

Synonyms: H-β-Ala-OBzl·p-tosylate, H-β-Ala-Obzl.TosOH, H-β-Ala-OBzl·TosOH

A18650.09 A18650.18
A18650.09EA 1210 CZK
A18650.09 A18650.18
β-Alanine benzyl ester p-toluenesulphonate 98%
β-Alanine benzyl ester p-toluenesulphonate

Purity: 98% CAS No.: 27019-47-2 Molecular Formula: C6H5CH2O2CCH2CH2NH2.HO3SC6H4CH3 Formula Weight: 351.42 Storage Temperature: 2°C to 8°C Melting Point: 138-139° MDL No.: MFCD00039061

Vzorec: C₁₀H₁₃NO₂
MW: 351,42 g/mol
Bod tání: 138…139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039061
CAS číslo: 27019-47-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR