Tisk…

4-Methoxytrityl alcohol 94%

Dodavatel: Thermo Fisher Scientific

Synonyms: (4-Methoxyphenyl)diphenylmethanol

A18620.09 A18620.18 A18620.30
A18620.09EA 1280 CZK
A18620.09 A18620.18 A18620.30
4-Methoxytrityl alcohol 94%
4-Methoxytrityl alcohol

Purity: 94% CAS No.: 847-83-6 Molecular Formula: C20H18O2 Formula Weight: 290.36 MDL No.: MFCD00087962

Vzorec: C₂₀H₁₈O₂
MW: 290.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00087962
CAS číslo: 847-83-6
EINECS: 212-693-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR