Sodium ((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate 99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
U 5′-P, Disodium uridine-5-monophosphate, Sodium ((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate, UMP, 5′-UMP-Na2, di-Sodium uridine-5-monophosphate, 5'-UMP 2Na, 5'-Uridylic acid disodium salt, Uridylic acid disodium salt, 5'-UMP disodium salt, Disodium UMP
Purity: 99%
CAS No.: 3387-36-8
Molecular Formula: C9H11N2Na2O9P
Formula Weight: 368.15
Storage Temperature: 2°C to 8°C
Melting Point: ca 209° dec.
Beilstein Reference: 3582885
MDL No.: MFCD00006525
Vzorec:
C₉H₁₁N₂Na₂O₉P MW: 368.15 g/mol Bod tání: ∼ 209 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006525 CAS číslo: 3387-36-8 EINECS: 222-211-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...