Tisk…

Nitrofurazone ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 5-Nitrofurfural Semicarbazone, 2-((5-Nitrofuran-2-yl)methylene)hydrazinecarboxamide, 5-Nitro-2-Furaldehyde Semicarbazone

A18593.14 A18593.22 A18593.30
A18593.14EA 845 CZK
A18593.14 A18593.22 A18593.30
Nitrofurazone ≥98%
Nitrofurazone

Purity: 98+% CAS No.: 59-87-0 Molecular Formula: C6H6N4O4 Formula Weight: 198.14 Melting Point: ca 240° dec. Beilstein Reference: 86403 MDL No.: MFCD00003225

Vzorec: C₆H₆N₄O₄
MW: 198,14 g/mol
Bod tání: ∼ 240 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00003225
CAS číslo: 59-87-0
EINECS: 200-443-1
Merck Index: 14,6600

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR