Tisk…

2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine 97%

Dodavatel: Thermo Fisher Scientific
Varování

A18533.03 A18533.06
A18533.03EA 1530 CZK
A18533.03 A18533.06
2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine 97%
2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine

Purity: 97% CAS No.: 1513-69-5 Molecular Formula: C5H4F3N3O Formula Weight: 179.10 Melting Point: 288-292° Beilstein Reference: 3608100 MDL No.: MFCD00068093

Vzorec: C₅H₄F₃N₃O
Bod tání: 288…292 °C
Storage Temperature: Ambient
MDL Number: MFCD00068093
CAS číslo: 1513-69-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR