Tisk…

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: PFBHA.HCl

A18368.01 A18368.03 A18368.06
A18368.01EA 2140 CZK
A18368.01 A18368.03 A18368.06
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥99%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride

Purity: 99+% CAS No.: 57981-02-9 Molecular Formula: C7H4F5NO.HCl Formula Weight: 249.57 Melting Point: ca 215° subl. Beilstein Reference: 4031190 MDL No.: MFCD00012953

Vzorec: C₇H₄F₅NO
Bod tání: ∼ 215 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00012953
CAS číslo: 57981-02-9
EINECS: 261-057-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR