Tisk…

Ethyl red

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-(Diethylamino)azobenzene-2'-carboxylic Acid, Diethyl Red

A18367.09 A18367.18 A18367.30
A18367.09EA 1170 CZK
A18367.09 A18367.18 A18367.30
Ethyl red
Ethyl red

CAS No.: 76058-33-8 Molecular Formula: C17H19N3O2 Formula Weight: 297.36 Melting Point: ca 135° dec. Beilstein Reference: 3366056 MDL No.: MFCD00002427

Vzorec: C₁₇H₁₉N₃O₂
MW: 297,36 g/mol
Bod tání: ∼ 135 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00002427
CAS číslo: 76058-33-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR