Sodium taurocholate hydrate 96%
Dodavatel: Thermo Fisher Scientific
A18346.14
A18346.06
A18346.03
A18346.14EA
19770
CZK
A18346.14
A18346.06
A18346.03
Sodium taurocholate hydrate 96%
Sodium taurocholate hydrate
Purity: 97%
CAS No.: 345909-26-4
Molecular Formula: C26H44NNaO7S.xH2O
Formula Weight: 537.73(anhy)
Physical Form: Crystals
Appearance: White
Optical Rotation: +23.0° (c=3 in water)
MDL No.: MFCD00150819
Vzorec:
C₂₆H₄₆NNaO₈S Storage Temperature: Ambient |
MDL Number:
MFCD00150819 CAS číslo: 345909-26-4 EINECS: 205-653-7 Merck Index: 14,9075 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...