Tisk…

2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide 99%

Dodavatel: Thermo Scientific
Varování

A18294.03 A18294.06
A18294.03EA 1090 CZK
A18294.03 A18294.06
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide 99%
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide

Purity: 99% CAS No.: 19805-75-5 Molecular Formula: C9H16N2O Formula Weight: 168.24 Melting Point: 179-183° MDL No.: MFCD00041846

Vzorec: C₉H₁₆N₂O
MW: 168,24 g/mol
Bod tání: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00041846
CAS číslo: 19805-75-5
EINECS: 243-331-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR