5-Chloro-2,1,3-benzothiadiazole 98%
Dodavatel: Thermo Scientific
Synonyms:
5-Chlorobenzo-2,1,3-thiadiazole
A18286.03
A18286.06
A18286.03EA
2750
CZK
A18286.03
A18286.06
5-Chloro-2,1,3-benzothiadiazole 98%
5-Chloro-2,1,3-benzothiadiazole
Purity: 98%
CAS No.: 2207-32-1
Molecular Formula: C6H3ClN2S
Formula Weight: 170.62
Melting Point: 55-57°
MDL No.: MFCD00174269
Vzorec:
C₆H₃ClN₂S MW: 170,62 g/mol Bod tání: 55…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00174269 CAS číslo: 2207-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...