Tisk…

(S)-2-Hydroxy-3-phenylpropanoic acid 98%

Dodavatel: Thermo Scientific

Synonyms: L-β-Phenyllactic acid, (S)-(-)-3-Phenyllactic Acid, L-3-Phenyllactic Acid, L(-)-3-Phenyllactic acid, (S)-(-)-2-Hydroxy-3-phenylpropionic Acid, L-(-)-3-Phenyllactic acid

A18203.03 A18203.06 A18203.14
A18203.03EA 879 CZK
A18203.03 A18203.06 A18203.14
(S)-2-Hydroxy-3-phenylpropanoic acid 98%
(S)-2-Hydroxy-3-phenylpropanoic acid

Purity: 98% CAS No.: 20312-36-1 Molecular Formula: C9H10O3 Formula Weight: 166.18 Storage Temperature: 2°C to 8°C Melting Point: 122-126° Beilstein Reference: 2209792 Optical Rotation: -24° (c=1 in water) MDL No.: MFCD00004244

Vzorec: C₉H₁₀O₃
MW: 166,17 g/mol
Bod tání: 122…126 °C
MDL Number: MFCD00004244
CAS číslo: 20312-36-1
EINECS: 243-726-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR