Tisk…

N-(2-Cyanoethyl)glycine 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: N-(2-Cyanoethyl)aminoacetic Acid

A18199.06 A18199.14
A18199.06EA 948 CZK
A18199.06 A18199.14
N-(2-Cyanoethyl)glycine 98%
N-(2-Cyanoethyl)glycine

Purity: 98% CAS No.: 3088-42-4 Molecular Formula: NCCH2CH2NHCH2CO2H Formula Weight: 128.13 Melting Point: ca 195° dec. Beilstein Reference: 1756230 UN No.: UN3439 MDL No.: MFCD00066903

Vzorec: C₅H₈N₂O₂
MW: 128,13 g/mol
Bod tání: ∼ 195 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00066903
CAS číslo: 3088-42-4
EINECS: 221-418-1
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR