Tisk…

6-Nitropiperonyl alcohol ≥98%

Dodavatel: Thermo Fisher Scientific

A18140.06 A18140.14
A18140.06EA 1660 CZK
A18140.06 A18140.14
6-Nitropiperonyl alcohol ≥98%
6-Nitropiperonyl alcohol

Purity: 98+% CAS No.: 15341-08-9 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 121-125° Beilstein Reference: 13749 MDL No.: MFCD00005825

Vzorec: C₈H₇NO₅
Bod tání: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00005825
CAS číslo: 15341-08-9
EINECS: 239-376-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR