2-Naphthyl phenyl ketone 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Benzoylnaphthalene
A18121.03
A18121.06
A18121.03EA
1540
CZK
A18121.03
A18121.06
2-Naphthyl phenyl ketone 98%
2-Naphthyl phenyl ketone
Purity: 98%
CAS No.: 644-13-3
Molecular Formula: C17H12O
Formula Weight: 232.28
Melting Point: 80-82°
Boiling Point: 216-216°/7mm
MDL No.: MFCD00004106
Vzorec:
C₁₇H₁₂O MW: 232,28 g/mol Bod varu: 216…216 °C (7 mmHg) Bod tání: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004106 CAS číslo: 644-13-3 EINECS: 211-410-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...