Tisk…

Monosodium 5-sulphoisophthalate 95%

Dodavatel: Thermo Fisher Scientific
Varování

A18028.22 A18028.30 A18028.36
A18028.22EA 1920 CZK
A18028.22 A18028.30 A18028.36
Monosodium 5-sulphoisophthalate 95%
Monosodium 5-sulphoisophthalate

Purity: 95% CAS No.: 6362-79-4 Molecular Formula: C8H5NaO7S Formula Weight: 268.18 Melting Point: 300° Beilstein Reference: 4170770 MDL No.: MFCD00007495

Vzorec: C₈H₆O₇S
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007495
CAS číslo: 6362-79-4
EINECS: 228-845-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR