Tisk…
Amberlyst® A-21 ion exchange resin
Dodavatel: Thermo Fisher Scientific
Synonyms: Amberlyst® A21
A17956.30 A17956.0B
A17956.30EA 1520 CZK
A17956.30 A17956.0B
Amberlyst® A-21 ion exchange resin
Amberlyst® A-21
CAS No.: 9049-93-8 Odour: Odourless MDL No.: MFCD00145842
Storage Temperature: Ambient
MDL Number: MFCD00145842
CAS číslo: 9049-93-8