Manganese(III) phosphate hydrate 99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A17868.09
A17868.18
A17868.09EA
1120
CZK
A17868.09
A17868.18
Manganese(III) phosphate hydrate 99%
Manganese(III) phosphate hydrate
Purity: 99%
CAS No.: 104663-56-1
Molecular Formula: MnPO4.xH2O
Formula Weight: 149.91(anhy)
MDL No.: MFCD00209658
Vzorec:
MnPO₄·xH₂O MW: 149,91 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00209658 CAS číslo: 104663-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...