Tisk…

L-Isoleucine methyl ester hydrochloride ≥98%

Dodavatel: Thermo Fisher Scientific

A17847.03 A17847.06 A17847.14
A17847.03EA 807 CZK
A17847.03 A17847.06 A17847.14
L-Isoleucine methyl ester hydrochloride ≥98%
L-Isoleucine methyl ester hydrochloride

Purity: 98+% CAS No.: 18598-74-8 Molecular Formula: C7H15NO2.HCl Formula Weight: 181.66 Melting Point: 98-100° Beilstein Reference: 3595132 MDL No.: MFCD00038911

Vzorec: C₇H₁₅NO₂
Bod tání: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00038911
CAS číslo: 18598-74-8
EINECS: 242-437-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR