Tisk…

Amberlite® IRA-410 (Cl) ion exchange resin

Dodavatel: Thermo Fisher Scientific

Synonyms: Amberlite® IRA-410 Cl type

A17734.0E A17734.22 A17734.36
A17734.0EEA 10020 CZK
A17734.0E A17734.22 A17734.36
Amberlite® IRA-410 (Cl) ion exchange resin
Amberlite® IRA-410 (Cl)

CAS No.: 9002-26-0 Odour: Amine-like MDL No.: MFCD00132710

Storage Temperature: Ambient
MDL Number: MFCD00132710
CAS číslo: 9002-26-0
Merck Index: 14,382

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR