8-Hydroxyjulolidine 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1,2,3,5,6,7-Hexahydropyrido[3,2,1-ij]quinolin-8-ol, 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol
A17692.03
A17692.06
A17692.03EA
2220
CZK
A17692.03
A17692.06
8-Hydroxyjulolidine 97%
8-Hydroxyjulolidine
Purity: 97%
CAS No.: 41175-50-2
Molecular Formula: C12H15NO
Formula Weight: 189.26
Melting Point: 132-135°
Beilstein Reference: 1530423
MDL No.: MFCD00006918
Vzorec:
C₁₂H₁₅NO MW: 189,26 g/mol Bod tání: 132…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006918 CAS číslo: 41175-50-2 EINECS: 255-247-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...