Tisk…

Nonenylsuccinic anhydride (mixture of branched chain isomers) 96%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: NSA, Isononenylsuccinic anhydride

A17607.30 A17607.18
A17607.30EA 4190 CZK
A17607.30 A17607.18
Nonenylsuccinic anhydride (mixture of branched chain isomers) 96%
Nonenylsuccinic anhydride (mixture of branched chain isomers)

Purity: 96% CAS No.: 28928-97-4 Molecular Formula: C13H20O3 Formula Weight: 224.30 Flash Point: 146°(295°F) Boiling Point: 126-130°/0.15mm MDL No.: MFCD00014552

Vzorec: C₁₃H₂₀O₃
MW: 224.30 g/mol
Bod varu: 126…130 °C (0.15 mmHg)
Hustota: 1.032
Bod vzplanutí: 146 °C (295 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014552
CAS číslo: 28928-97-4
EINECS: 249-317-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR