N-Carbobenzoxy-L-threonine 99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Z-Thr-OH, (2S,3R)-2-(((Benzyloxy)carbonyl)amino)-3-hydroxybutanoic acid, N-Cbz-L-threonine
A17565.06
A17565.14
A17565.06EA
1050
CZK
A17565.06
A17565.14
N-Carbobenzoxy-L-threonine 99%
N-Carbobenzoxy-L-threonine
Purity: 99%
CAS No.: 19728-63-3
Molecular Formula: C12H15NO5
Formula Weight: 253.26
Melting Point: 103-105°
Beilstein Reference: 2335409
Optical Rotation: -4.5° (c=4 in acetic acid)
MDL No.: MFCD00065948
Vzorec:
C₁₂H₁₅NO₅ MW: 253.25 g/mol Bod tání: 103…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065948 CAS číslo: 19728-63-3 EINECS: 243-258-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...