Amberlyst® A-26 (OH) ion exchange resin
Dodavatel: Thermo Fisher Scientific
A17361.30
A17361.0B
A17361.30EA
2840
CZK
A17361.30
A17361.0B
Amberlyst® A-26 (OH) ion exchange resin
Amberlyst® A-26 (OH)
CAS No.: 39339-85-0
MDL No.: MFCD00145579
Storage Temperature:
Ambient |
MDL Number:
MFCD00145579 CAS číslo: 39339-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...