Cyclohexylamine hydrochloride ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Cyclohexylamine Hydrochloride
A17308.22
A17308.36
A17308.22EA
930
CZK
A17308.22
A17308.36
Cyclohexylamine hydrochloride ≥98%
Cyclohexylamine hydrochloride
Purity: 98+%
CAS No.: 4998-76-9
Molecular Formula: C6H13N.HCl
Formula Weight: 135.64
Odour: Odourless
Melting Point: 206-209°
Beilstein Reference: 3613002
MDL No.: MFCD00050540
Vzorec:
C₆H₁₃N MW: 135,64 g/mol Bod tání: 206…209 °C Storage Temperature: Ambient |
MDL Number:
MFCD00050540 CAS číslo: 4998-76-9 EINECS: 225-661-4 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...