N-Acetyl-L-tyrosine 99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
N-Acetyl-L-Tyrosine, (S)-2-Acetamido-3-(4-hydroxyphenyl)propanoic acid, Ac-Tyr-OH
A17307.09
A17307.18
A17307.30
A17307.09EA
2130
CZK
A17307.09
A17307.18
A17307.30
N-Acetyl-L-tyrosine 99%
N-Acetyl-L-tyrosine
Purity: 99%
CAS No.: 537-55-3
Molecular Formula: C11H13NO4
Formula Weight: 223.23
Melting Point: 150-151°
Beilstein Reference: 2697172
Optical Rotation: +58° (c=5 in ethanol)
MDL No.: MFCD00037190
Vzorec:
C₁₁H₁₃NO₄ MW: 223.23 g/mol Bod tání: 150…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037190 CAS číslo: 537-55-3 EINECS: 208-671-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...