L-Glutamic acid monopotassium salt monohydrate 97
Dodavatel: Thermo Fisher Scientific
A17232.30
A17232.0B
A17232.30EA
1770
CZK
A17232.30
A17232.0B
L-Glutamic acid monopotassium salt monohydrate 97
L-Glutamic acid monopotassium salt monohydrate
Purity: 97+%
CAS No.: 6382-01-0
Molecular Formula: C5H8KNO4.H2O
Formula Weight: 203.25 (185.23anhy)
Melting Point: 115-118°
MDL No.: MFCD00150343
Vzorec:
C₅H₈KNO₄·H₂O MW: 203.25 g/mol (185.23 anhy) Bod tání: 115…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150343 CAS číslo: 6382-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...