Xanthotoxin 99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A17108.03
A17108.06
A17108.14
A17108.03EA
2050
CZK
A17108.03
A17108.06
A17108.14
Xanthotoxin 99%
Xanthotoxin
Purity: 99%
CAS No.: 298-81-7
Molecular Formula: C12H8O4
Formula Weight: 216.20
Melting Point: 146-150°
Beilstein Reference: 196453
MDL No.: MFCD00005009
Vzorec:
C₁₂H₈O₄ Bod tání: 146…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005009 CAS číslo: 298-81-7 EINECS: 206-066-9 Merck Index: 14,5988 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...