3-Indoxyl sulphate potassium salt 97%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Potassium 1H-indol-3-yl sulphate, 3-Indolyl sulphate potassium salt
A17079.03
A17079.01
A17079.06
A17079.03EA
6700
CZK
A17079.03
A17079.01
A17079.06
3-Indoxyl sulphate potassium salt 97%
3-Indoxyl sulphate potassium salt
Purity: 97%
CAS No.: 2642-37-7
Molecular Formula: C8H6KNO4S
Formula Weight: 251.31
Storage Temperature: -30°C to -10°C
Melting Point: 165° dec.
Beilstein Reference: 3921324
MDL No.: MFCD00037931
Vzorec:
C₈H₆KNO₄S MW: 251.31 g/mol Bod tání: 165 °C (rozkl.) |
MDL Number:
MFCD00037931 CAS číslo: 2642-37-7 EINECS: 220-145-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...