Tisk…

1-Phenylpiperazine hydrochloride 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A16995.14 A16995.22
A16995.14EA 2000 CZK
A16995.14 A16995.22
1-Phenylpiperazine hydrochloride 99%
1-Phenylpiperazine hydrochloride

Purity: 99% CAS No.: 2210-93-7 Molecular Formula: C10H14N2.HCl Formula Weight: 198.70 Odour: Odourless Melting Point: 247-250° UN No.: UN2811 MDL No.: MFCD00012758

Vzorec: C₁₀H₁₄N₂·HCl
MW: 198.70 g/mol
Bod tání: 247…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012758
CAS číslo: 2210-93-7
EINECS: 218-646-9
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR