Tisk…

L-Lysine ethyl ester dihydrochloride 99%

Dodavatel: Thermo Fisher Scientific
Varování

A16824.14 A16824.22
A16824.14EA 2490 CZK
A16824.14 A16824.22
L-Lysine ethyl ester dihydrochloride 99%
L-Lysine ethyl ester dihydrochloride

Purity: 99% CAS No.: 3844-53-9 Molecular Formula: C8H18N2O2.2HCl Formula Weight: 247.17 Melting Point: ca 145° dec. Beilstein Reference: 3697099 MDL No.: MFCD00039068

Vzorec: C₈H₂₀Cl₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00039068
CAS číslo: 3844-53-9
EINECS: 223-340-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR