Tisk…

Evans blue

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A16774.09 A16774.18
A16774.09EA 1950 CZK
A16774.09 A16774.18
Evans blue
Evans blue

CAS No.: 314-13-6 Molecular Formula: C34H24N6Na4O14S4 Formula Weight: 960.82 MDL No.: MFCD00004021


Applications: Direct Blue 53. Inhibits L-glutamate uptake, and AMPA and kainate receptor currents

Vzorec: C₃₄H₂₈N₆O₁₄S₄
Storage Temperature: Ambient
MDL Number: MFCD00004021
CAS číslo: 314-13-6
EINECS: 206-242-5
Merck Index: 14,3905

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR