Tisk…
4,4'-Diaminostilbene-2,2'-disulphonic Acid 95%
Dodavatel: Thermo Fisher Scientific
Synonyms: Amsonic acid, 4,4-Diamino-2,2-stilbenedisulphonic acid
A16664.14 A16664.22
A16664.14EA 1390 CZK
A16664.14 A16664.22
4,4'-Diaminostilbene-2,2'-disulphonic Acid 95%
4,4'-Diaminostilbene-2,2'-disulphonic Acid
Purity: 95% CAS No.: 81-11-8 Molecular Formula: C14H14N2O6S2 Formula Weight: 370.41 Melting Point: 300° Beilstein Reference: 629516 MDL No.: MFCD00024946
Vzorec: C₁₄H₁₄N₂O₆S₂
MDL Number: MFCD00024946
CAS číslo: 81-11-8
Merck Index: 14,595