Tisk…

3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A16317.06 A16317.14 A16317.22
A16317.06EA 1580 CZK
A16317.06 A16317.14 A16317.22
3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate 97%
3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate

Purity: 97% CAS No.: 149022-15-1 Molecular Formula: C8H9N3S.HCl.xH2O Formula Weight: 215.71(anhy) Melting Point: ca 275° dec. Beilstein Reference: 3571041 UN No.: UN2811 MDL No.: MFCD00149370

Vzorec: C₈H₉N₃S
Storage Temperature: Ambient
MDL Number: MFCD00149370
CAS číslo: 149022-15-1
EINECS: 238-428-7
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR