Tisk…
Calcium glycerophosphate hydrate 97%
Dodavatel: Thermo Fisher Scientific
Synonyms: Glycerophosphoric acid calcium salt hydrate
A16269.30 A16269.0B
A16269.30EA 2020 CZK
A16269.30 A16269.0B
Calcium glycerophosphate hydrate 97%
Calcium glycerophosphate hydrate
Purity: 97% CAS No.: 28917-82-0 Molecular Formula: C3H7CaO6P.xH2O Formula Weight: 210.15(anhy) MDL No.: MFCD00151656
Vzorec: C₃H₇CaO₆P·xH₂O
MW: 210.15 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00151656
CAS číslo: 28917-82-0
EINECS: 249-312-0
Merck Index: 14,1670