N-Boc-L-serine, (max. 10% H₂O) 98% (dry weight)
Dodavatel: Thermo Scientific
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid, Boc-Ser-OH, N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine, N-(tert-Butoxycarbonyl)-L-serine hydrate, BOC-L-Serine hydrate
A16224.06
A16224.14
A16224.06EA
1860
CZK
A16224.06
A16224.14
N-Boc-L-serine, (max. 10% H₂O) 98% (dry weight)
N-Boc-L-serine
Purity: 98%
CAS No.: 3262-72-4
Molecular Formula: C8H15NO5
Formula Weight: 205.21
Storage Temperature: 2°C to 8°C
Melting Point: 90-91° dec.
Beilstein Reference: 2212252
Optical Rotation: -3.5° (c=2 in acetic acid)
MDL No.: MFCD00037243
Vzorec:
C₈H₁₅NO₅ MW: 205,21 g/mol Bod tání: 90…91 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00037243 CAS číslo: 3262-72-4 EINECS: 221-867-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...