Tisk…

Formaldehyde sodium bisulphite 95%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Sodium Hydroxymethanesulphonate, Hydroxymethanesulphonic acid sodium salt

A16194.36 A16194.0E A16194.22
A16194.36EA 2830 CZK
A16194.36 A16194.0E A16194.22
Formaldehyde sodium bisulphite 95%
Formaldehyde sodium bisulphite

Purity: 95% CAS No.: 870-72-4 Molecular Formula: HOCH2SO3Na Formula Weight: 134.09 Odour: Odourless Melting Point: 200° dec. MDL No.: MFCD00044664

Vzorec: CH₄O₄S
Storage Temperature: Ambient
MDL Number: MFCD00044664
CAS číslo: 870-72-4
EINECS: 212-800-9
Merck Index: 14,4236

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR