Tisk…

1,3-Bis(hydroxymethyl)urea 90%, tech.

Dodavatel: Thermo Fisher Scientific

A16060.0E A16060.22 A16060.36
A16060.0EEA 10060 CZK
A16060.0E A16060.22 A16060.36
1,3-Bis(hydroxymethyl)urea 90%, tech.
1,3-Bis(hydroxymethyl)urea

Purity: 90% CAS No.: 140-95-4 Molecular Formula: C3H8N2O3 Formula Weight: 120.11 Odour: Odourless pH: 7-8 Melting Point: 124-128° MDL No.: MFCD00014414

Vzorec: C₃H₈N₂O₃
Bod tání: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00014414
CAS číslo: 140-95-4
EINECS: 205-444-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR