Tisk…

Nε-carbobenzoxy-L-lysine 98%

Dodavatel: Thermo Scientific

Synonyms: Nε-Cbz-L-lysine, (S)-2-Amino-6-(((benzyloxy)carbonyl)amino)hexanoic acid, H-Lys(Z)-OH

A16022.06 A16022.14
A16022.06EA 1500 CZK
A16022.06 A16022.14
Nε-carbobenzoxy-L-lysine 98%
Nepsilon-carbobenzoxy-L-lysine

Purity: 98% CAS No.: 1155-64-2 Molecular Formula: C14H20N2O4 Formula Weight: 280.32 Melting Point: 259° dec. Beilstein Reference: 2222482 MDL No.: MFCD00002638

Vzorec: C₁₄H₂₀N₂O₄
MW: 280,32 g/mol
Bod tání: 259 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00002638
CAS číslo: 1155-64-2
EINECS: 214-585-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR