Tisk…

3-(4-Methoxyphenyl)-3-oxopropanenitrile 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-Methoxybenzoylacetonitrile

A15973.06 A15973.14
A15973.06EA 3260 CZK
A15973.06 A15973.14
3-(4-Methoxyphenyl)-3-oxopropanenitrile 98%
3-(4-Methoxyphenyl)-3-oxopropanenitrile

Purity: 98% CAS No.: 3672-47-7 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: 125-130° Beilstein Reference: 743909 MDL No.: MFCD00067889

Vzorec: C₁₀H₉NO₂
MW: 175,19 g/mol
Bod tání: 125…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00067889
CAS číslo: 3672-47-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR