Tisk…

Diacetone acrylamide 99%

Dodavatel: Thermo Scientific
Varování

Synonyms: N-(2-Methyl-4-oxopentan-2-yl)acrylamide, N-(1,1-Dimethyl-3-oxobutyl)acrylamide

A15940.0B A15940.18 A15940.30
A15940.0BEA 11530 CZK
A15940.0B A15940.18 A15940.30
Diacetone acrylamide 99%
Diacetone acrylamide

Purity: 99% CAS No.: 2873-97-4 Molecular Formula: CH3COCH2C(CH3)2NHCOCH=CH2 Formula Weight: 169.23 Flash Point: 110°(230°F) Melting Point: 54-56° Beilstein Reference: 1928444 MDL No.: MFCD00008788

Vzorec: C₉H₁₅NO₂
MW: 169,22 g/mol
Bod tání: 54…56 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008788
CAS číslo: 2873-97-4
EINECS: 220-713-2
Merck Index: 14,2963

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR