Tisk…

5-Chloro-6-hydroxynicotinic acid 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-Chloro-6-hydroxy-3-pyridinecarboxylic Acid

A15737.06 A15737.14
A15737.06EA 841 CZK
A15737.06 A15737.14
5-Chloro-6-hydroxynicotinic acid 98%
5-Chloro-6-hydroxynicotinic acid

Purity: 98% CAS No.: 54127-63-8 Molecular Formula: C6H4ClNO3 Formula Weight: 173.56 Melting Point: 300° MDL No.: MFCD00052950

Vzorec: C₆H₄ClNO₃
MW: 173,55 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00052950
CAS číslo: 54127-63-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR