D-(+)-Glucosamine hydrochloride ≥98%
Dodavatel: Thermo Scientific
Synonyms:
2-Amino-2-deoxy-D-glucopyranose Hydrochloride, D-Glucosamine hydrochloride, (2R,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride
A15532.0B
A15532.18
A15532.30
A15532.0BEA
11530
CZK
A15532.0B
A15532.18
A15532.30
D-(+)-Glucosamine hydrochloride ≥98%
D-(+)-Glucosamine hydrochloride
Purity: 98+%
CAS No.: 66-84-2
Molecular Formula: C6H13NO5.HCl
Formula Weight: 215.64
Melting Point: ca 192° dec.
Beilstein Reference: 4157370
Optical Rotation: +71.5° (c=1 in water, 20h)
MDL No.: MFCD00067674
Vzorec:
C₆H₁₃NO₅ MW: 215,63 g/mol Bod tání: ∼ 192 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00135831 CAS číslo: 66-84-2 EINECS: 200-638-1 Merck Index: 14,4458 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...