Tisk…

Metaldehyde 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Metacetaldehyde, Aldehyde (meta), Acetaldehyde Tetramer

A15292.18 A15292.0B A15292.30
A15292.18EA 1090 CZK
A15292.18 A15292.0B A15292.30
Metaldehyde 98%
Metaldehyde

Purity: 98% CAS No.: 108-62-3 Molecular Formula: C8H16O4 Formula Weight: 176.22 Flash Point: 55°(131°F) Melting Point: 192-196° subl. UN No.: UN1332 MDL No.: MFCD00071549

Vzorec: C₈H₁₆O₄
MW: 176.21 g/mol
Bod tání: 192…196 °C (subl.)
Bod vzplanutí: 55 °C (131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134115
CAS číslo: 108-62-3
EINECS: 203-600-2
UN: 1332
ADR: 4.1 III
Merck Index: 14,5923

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR