3-Cyclohexene-1-carboxylic acid 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Cyclohex-3-enecarboxylic acid, 3-Cyclohexenecarboxylic acid, 1,2,3,6-Tetrahydrobenzoic Acid
A15229.06
A15229.14
A15229.06EA
2390
CZK
A15229.06
A15229.14
3-Cyclohexene-1-carboxylic acid 98%
3-Cyclohexene-1-carboxylic acid
Purity: 98%
CAS No.: 4771-80-6
Molecular Formula: C7H10O2
Formula Weight: 126.16
Flash Point:
Vzorec:
C₇H₁₀O₂ MW: 126,16 g/mol Bod varu: 120…122 °C (10 mmHg) Bod tání: 16…18 °C Hustota: 1,082 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013781 CAS číslo: 4771-80-6 EINECS: 225-314-7 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...