Tisk…

3,5-Di-tert-butyl-4-hydroxyphenylacetic acid 98%

Dodavatel: Thermo Scientific

A15015.03 A15015.06
A15015.03EA 1220 CZK
A15015.03 A15015.06
3,5-Di-tert-butyl-4-hydroxyphenylacetic acid 98%
3,5-Di-tert-butyl-4-hydroxyphenylacetic acid

Purity: 98% CAS No.: 1611-03-6 Molecular Formula: C16H24O3 Formula Weight: 264.37 Melting Point: 154-158° MDL No.: MFCD00156138

Vzorec: C₁₆H₂₄O₃
MW: 264,37 g/mol
Bod tání: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00156138
CAS číslo: 1611-03-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR